Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-hexenoate, 98%

Catalog No. AAA1973414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1973414 25 g
AAA1973406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1973414 Supplier Thermo Scientific Chemicals Supplier No. A1973414
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2396-83-0 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-83-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00036524
InChI Key VTSFIPHRNAESED-AATRIKPKSA-N
Synonym ethyl 3-hexenoate, 3-hexenoic acid, ethyl ester, ethyl hex-3-enoate, ethyl e-hex-3-enoate, 3-hexenoic acid, ethyl ester, e, ethyl trans-3-hexenoate, ethyl e-3-hexenoate, e-3-hexenoic acid ethyl ester, ethyl hydrosorbate, 3-hexenoic acid, ethyl ester, 3e
PubChem CID 5362622
IUPAC Name ethyl (E)-hex-3-enoate
SMILES CCOC(=O)C\C=C\CC

Specifications

Density 0.89
Boiling Point 63°C to 64°C (12 mmHg)
Flash Point 59°C (139°F)
Refractive Index 1.426
Quantity 25 g
UN Number UN3272
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl 3-hexenoate
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 219-257-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.