Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Iodobenzoate 98.0+%, TCI America™
SDP

Catalog No. I053625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I053625G 25 g
I05365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I053625G Supplier TCI America Supplier No. I053625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58313-23-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD00044769
InChI Key POGCXCWRMMXDAQ-UHFFFAOYSA-N
Synonym ethyl-3-iodobenzoate, 3-iodobenzoic acid ethyl ester, benzoic acid, 3-iodo-, ethyl ester, ethyl m-iodobenzoate, m-iodobenzoic acid ethyl ester, pubchem3883, ethyl-3-iodo-benzoate, acmc-1aupw, ethyl 3-iodobenzoate, rarechem al bi 0709
PubChem CID 143542
IUPAC Name ethyl 3-iodobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)I

Specifications

Color Yellow
Boiling Point 161°C
Quantity 25 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Iodobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.