Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (3-Methoxybenzoyl)acetate 98.0+%, TCI America™
SDP

Catalog No. E11211G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11211G 1 g
E11215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11211G Supplier TCI America Supplier No. E11211G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27834-99-7
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00086704
InChI Key FDPPVAYPZOORBP-UHFFFAOYSA-N
Synonym (3-Methoxybenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(3-Methoxyphenyl)-3-oxopropionate, 3-(3-Methoxyphenyl)-3-oxopropionic Acid Ethyl Ester
PubChem CID 2759696
IUPAC Name ethyl 3-(3-methoxyphenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(OC)=CC=C1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 1 g
Formula Weight 222.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (3-Methoxybenzoyl)acetate
EINECSNumber (3)-1745
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.