Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 3-methyl-2-oxobutyrate

Catalog No. 2184565G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-845-65G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-845-65G Supplier Sigma Aldrich Supplier No. 2184565G
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyl 3-methyl-2-oxobutyrate

Specifications

CAS 20201-24-5
Density 0.989 g/mL (at 25°C (literature))
Boiling Point 62°C (11 mmHg)
Molecular Formula C7H12O3
Linear Formula (CH3)2 CHCOCOOCH2CH3
Refractive Index n20/D 1.410 (literature)
MDL Number MFCD00009122
Quantity 250 mg
Synonym Ethyl dimethylpyruvate; Ketovaline ethyl ester
Molecular Weight (g/mol) 144.17
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.