Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-Methylcrotonate 97.0+%, TCI America™
SDP

Catalog No. M144125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M144125ML 25 mL
M14415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M144125ML Supplier TCI America Supplier No. M144125ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 638-10-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00009187
InChI Key UTXVCHVLDOLVPC-UHFFFAOYSA-N
Synonym 3,3-Dimethylacrylic Acid Ethyl Ester, Ethyl 3,3-Dimethylacrylate, Ethyl 3-Methyl-2-butenoate, 3-Methylcrotonic Acid Ethyl Ester, 3-Methyl-2-butenoic Acid Ethyl Ester
PubChem CID 12516
IUPAC Name ethyl 3-methylbut-2-enoate
SMILES CCOC(=O)C=C(C)C

Specifications

Color Colorless
Quantity 25 mL
UN Number 3272
Formula Weight 128.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 3-Methylcrotonate
RTECSNumber GQ5750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.