Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-(1-Piperazinyl)benzoate 98.0+%, TCI America™
SDP

Catalog No. E126025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E126025G 25 g
E12605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E126025G Supplier TCI America Supplier No. E126025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80518-57-6
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD04973340
InChI Key OQEHTFFLOHTFSB-UHFFFAOYSA-N
Synonym ethyl 4-piperazin-1-yl benzoate, ethyl 4-1-piperazinyl benzoate, 4-piperazin-1-yl-benzoic acid ethyl ester, 1-4-ethoxycarbonylphenyl piperazine, ethyl 4-1-piperazino benzoate, ethyl 4-piperazin-1yl benzoate, benzoic acid, 4-1-piperazinyl-, ethyl ester, 4-piperazin-1-yl benzoic acidethylester, 4-1-piperazinyl benzoic acid ethyl ester
PubChem CID 2761178
IUPAC Name ethyl 4-(piperazin-1-yl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 102°C
Color Yellow
Quantity 25 g
Formula Weight 234.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-(1-Piperazinyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.