Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4,4,4-Trichloroacetoacetate 97.0+%, TCI America™
SDP

Catalog No. T128510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T128510G 10 g
T128525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T128510G Supplier TCI America Supplier No. T128510G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3702-98-5
Molecular Formula C6H7Cl3O3
Molecular Weight (g/mol) 233.469
MDL Number MFCD00043329
InChI Key XGIRWPYENXRJMZ-UHFFFAOYSA-N
Synonym 4,4,4-Trichloroacetoacetic Acid Ethyl Ester
PubChem CID 77294
IUPAC Name ethyl 4,4,4-trichloro-3-oxobutanoate
SMILES CCOC(=O)CC(=O)C(Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 234°C
Quantity 10 g
Formula Weight 233.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-Trichloroacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.