Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-dimethylaminobenzoate, 99+%

Catalog No. AC118401000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118401000 100 g Plastic bottle
AC118400050 5 g Glass Bottle
AC118405000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118401000 Supplier Thermo Scientific Chemicals Supplier No. 118401000
Only null left
Add to Cart

CAS: 10287-53-3 | C11H15NO2 | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10287-53-3
MDL Number MFCD00009115
InChI Key FZUGPQWGEGAKET-UHFFFAOYSA-N
Synonym ethyl 4-dimethylamino benzoate, parbenate, benzoic acid, 4-dimethylamino-, ethyl ester, kayacure epa, ethyl-p-dimethylaminobenzoate, ethyl-4-dimethylaminobenzoate, n,n-dimethylbenzocaine, speedcure edb, ethyl p-dimethylaminobenzoate, unii-829s8d3y0x
PubChem CID 25127
ChEBI CHEBI:52073
IUPAC Name ethyl 4-(dimethylamino)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 62°C to 65°C
Color White to Yellow
Density 1.0600g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C11H15NO2
Linear Formula (CH3)2NC6H4CO2C2H5
Quantity 100 g
Beilstein 14, I, 571
Molecular Weight (g/mol) 193.246
Formula Weight 193.25
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 4-dimethylaminobenzoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.