Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-chlorophenylacetate, 98+%

Catalog No. AC388840050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388840050 5 g Glass bottle
AC388840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC388840050 Supplier Thermo Scientific Chemicals Supplier No. 388840050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14062-24-9 | C10H11ClO2 | 198.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14062-24-9
MDL Number MFCD00018791
InChI Key UTWBWFXECVFDPZ-UHFFFAOYSA-N
Synonym ethyl 4-chlorophenylacetate, ethyl 4-chlorophenyl acetate, ethyl 2-4-chlorophenyl acetate, 4-chlorophenyl acetic acid ethyl ester, ethyl p-chlorophenylacetate, benzeneacetic acid, 4-chloro-, ethyl ester, 4-chloro-phenyl-acetic acid ethyl ester, ethyl 4-chloro-phenylacetate, ksc522e5p
PubChem CID 84177
IUPAC Name ethyl 2-(4-chlorophenyl)acetate
SMILES CCOC(=O)CC1=CC=C(C=C1)Cl

Specifications

Color Colorless
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Low Melting Solid
Assay Percent Range 98+%
Molecular Formula C10H11ClO2
Linear Formula ClC6H4CH2C(O)OCH2CH3
Molecular Weight (g/mol) 198.646
Formula Weight 198.65
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-chlorophenylacetate
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.