Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (4-Fluorobenzoyl)acetate 98.0+%, TCI America™
SDP

Catalog No. F043525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F043525G 25 g
F04351G 1 g
F04355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F043525G Supplier TCI America Supplier No. F043525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1999-00-4
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD03093631
InChI Key SJUXLKYJKQBZLM-UHFFFAOYSA-N
Synonym ethyl 3-4-fluorophenyl-3-oxopropanoate, ethyl 4-fluorobenzoylacetate, ethyl 4-fluorobenzoyl acetate, 4-fluorobenzoylacetic acid ethyl ester, ethyl p-fluorobenzoyl acetate, 4-fluorobenzoyl acetic acid ethyl ester, ethyl 3-4-fluorophenyl-3-oxopropionate, ethyl 3-4'-fluorophenyl-3-oxopropanoate, ethyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid ethyl ester
PubChem CID 2758844
IUPAC Name ethyl 3-(4-fluorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC=C(C=C1)F

Specifications

Color Colorless
Boiling Point 120°C
Quantity 25 g
Formula Weight 210.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl (4-Fluorobenzoyl)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.