Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-Methylthiazole-5-carboxylate 97.0+%, TCI America™
SDP

Catalog No. E11821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11821G 1 g
E11825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11821G Supplier TCI America Supplier No. E11821G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20582-55-2
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00626873
InChI Key WISQBJLUORKXNY-UHFFFAOYSA-N
Synonym ethyl 4-methylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 4-methyl-, ethyl ester, 4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 4-methyl-ethylester, 4-methyl-thiazole-5-carboxylic acid ethyl ester, 4-methylthiazole-5-carboxylic acid ethyl ester, ethyl4-methyl-5-thiazoleactate, ethylmethylthiazolecarboxylate, 4-methyl-5-ethoxycarbonyl thiazole, 4-methylthiazol-5-carbonsaureathylester
PubChem CID 298610
IUPAC Name ethyl 4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=CS1)C

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 233°C
Quantity 1 g
Formula Weight 171.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl 4-Methylthiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.