Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-piperidone-3-carboxylate hydrochloride, 97%

Catalog No. AAA1213122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1213122 100 g
AAA1213106 5 g
AAA1213114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1213122 Supplier Thermo Scientific Chemicals Supplier No. A1213122
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4644-61-5 | C8H14ClNO3 | 207.654 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4644-61-5
Molecular Formula C8H14ClNO3
Molecular Weight (g/mol) 207.654
MDL Number MFCD00043267
InChI Key RTYXWLMQWSFXNI-UHFFFAOYSA-N
Synonym ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
PubChem CID 107315
IUPAC Name ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point ∼172°C (decomposition)
Quantity 100 g
Beilstein 3711279
Formula Weight 207.66
Percent Purity 97%
Chemical Name or Material Ethyl 4-piperidone-3-carboxylate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-073-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.