accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3COONHO

CAS RN 4644-61-5

IUPAC Name: ethyl 4-oxopiperidine-3-carboxylate hydrochloridyl
Synonyms: ethyl 4-oxopiperidine-3-carboxylate hydrochloridyl
Molecular Weight (g/mol): 206.65
Molecular Formula: C8H13ClNO3
InChi Key: OTNOTUGMYUCNQB-UHFFFAOYNA-N
SMILES: [Cl].CCOC(=O)C1CNCCC1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)