Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-Bromohexanoate 98.0+%, TCI America™
SDP

Catalog No. B1465250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1465250G 250 g
B146525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1465250G Supplier TCI America Supplier No. B1465250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Color Yellow
Boiling Point 130°C
Quantity 250 g
UN Number 3272
Formula Weight 223.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 6-Bromohexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.