Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate, 98%, Thermo Scientific™

Catalog No. AAH6443003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6443003 1 g
AAH6443006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6443003 Supplier Thermo Scientific Chemicals Supplier No. H6443003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59128-04-0
Molecular Formula C11H11BrN2O2
Molecular Weight (g/mol) 283.125
MDL Number MFCD16172180
InChI Key VBLCCEAJXYZCRL-UHFFFAOYSA-N
Synonym ethyl 2-6-bromoimidazo 1,2-a pyridin-2-yl acetate, ethyl 6-bromoimidazo 1,2-a pyridine-2-yl acetate, 6-bromo-imidazo 1,2-a pyridin-2-yl-acetic acid ethyl ester, imidazo 1,2-a pyridine-2-acetic acid, 6-bromo-, ethyl ester
PubChem CID 12269134
IUPAC Name ethyl 2-(6-bromoimidazo[1,2-a]pyridin-2-yl)acetate
SMILES CCOC(=O)CC1=CN2C=C(C=CC2=N1)Br

Specifications

Melting Point 55°C to 56°C
Color Gray
Quantity 1 g
Beilstein 791271
Formula Weight 283.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.