Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 6-hydroxyhexanoate

Catalog No. 37480610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-480-610G 10 g
37-480-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-480-610G Supplier Sigma Aldrich Supplier No. 37480610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl 6-hydroxyhexanoate is an ethyl ester
  • Its concentration in bordeaux red wines has been evaluated
  • Two-step preparation of ethyl 6-hydroxyhexanoate via hydrolysis of e-caprolactone has been reported.ethyl 6-hydroxyhexanoate is suitable for use in the synthesis of series of model phenol carbonate ester prodrugs having fatty acid-like structures
  • It may be used in the preparation of alkyl triflates
  • It may be used in the synthesis of ethyl 6-(trifluoromethylsulfonyloxy)hexanoate

Specifications

CAS 5299-60-5
Density 0.985 g/mL (at 25°C (literature))
Boiling Point 127°C to 128°C (12 mmHg)
Molecular Formula C8H16O3
Refractive Index n20/D 1.437 (literature)
Linear Formula HO(CH2)5 CO2C2H5
MDL Number MFCD00075601
Quantity 10 g
Molecular Weight (g/mol) 160.21
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.