Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 9-Bromononanoate 95.0+%, TCI America™
SDP

Catalog No. B23241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23241G 1 g
B23245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23241G Supplier TCI America Supplier No. B23241G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28598-81-4
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.191
MDL Number MFCD06797065
InChI Key KXZBPLOPBKIUTC-UHFFFAOYSA-N
Synonym 9-Bromononanoic Acid Ethyl Ester
PubChem CID 560317
IUPAC Name ethyl 9-bromononanoate
SMILES CCOC(=O)CCCCCCCCBr

Specifications

Color Yellow
Boiling Point 180°C
Quantity 1 g
Formula Weight 265.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 9-Bromononanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.