Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetate CHROMASOLV™, for HPLC, ≥99.7%, Solstice

Catalog No. 6000569 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-005-69 1 L
60-005-76 6 x 1 L
60-005-74 4 x 2.5 L
60-005-75 4 x 4 L
60-005-68 100 mL
60-005-72 2 L
60-005-70 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Repeated exposure may cause skin dryness or cracking.
Catalog No. 60-005-69 Supplier Solstice Supplier No. 348581L
Add to Cart
Add to Cart
Repeated exposure may cause skin dryness or cracking.

CHROMASOLV™, for HPLC,≥99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Density 0.902g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.3720
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
Beilstein 506104
Boiling Point 76.5°C to 77.5°C
Formula Weight 88.11g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV
Chemical Name or Material Ethyl acetate, for HPLC
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
EINECSNumber 205-500-4
RTECSNumber AH5425000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.