Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl acetate/Ethanol 31
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 745588
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This SOLUTIONon of Ethyl acetate/Ethanol is a greener alternative to dichloromethane for chromatographic purification and thus has been enhanced for Safer Solvents and Auxiliaries. Click here for more information.
Catalog No. 50-290-6848
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.