Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, NF, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602499 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
Packaging:
Amber Glass Bottle
Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-499 4 L Amber Glass Bottle
18-602-497 500 mL Amber Glass Bottle
18-602-498 6 x 500 mL Amber Glass Bottle
18-602-500 4 x 4 L Amber Glass Bottle
18-602-501 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-499 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ET1054LTGL
Add to Cart
Add to Cart

ET105, 141-78-6, C4H8O2

Spectrum™ Chemical Ethyl Acetate, NF is used as a diluent and solvent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
UN Number UN1173
Boiling Point 77°C
Percent Purity 98 to 102%
Grade NF
Chemical Name or Material Ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.