Learn More
Ethyl Acetate, NF, 98-102%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Ethyl Acetate, NF is used as a diluent and solvent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 4 x 4 L |
| Packaging | Amber Glass Bottle |
| UN Number | UN1173 |
| Boiling Point | 77°C |
| Percent Purity | 98 to 102% |
| Grade | NF |
| Chemical Name or Material | Ethyl Acetate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.