Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, puriss., ≥99.5% (GC); Solstice

Catalog No. 60045764 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
35 kg
4 x 2.5 L
6 x 1 L
180 kg
Packaging:
Aluminum Bottle
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-764 2.5 L Poly Bottle
60-045-769 6 x 1 L Poly Bottle
60-045-766 4 x 2.5 L Poly Bottle
60-045-765 35 kg Stainless Steel Can
60-045-761 180 kg Metal Drum
60-045-763 1 L Glass Bottle
60-045-762 1 L Poly Bottle
60-045-768 5 L Aluminum Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 60-045-764 Supplier Solstice Supplier No. 272272.5L
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99.5% (GC)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C
Quantity 2.5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
UN Number UN1173
Beilstein 506104
Boiling Point 76.5°C to 77.5°C
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Ethyl Acetate, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Ethyl acetate
EINECSNumber 205-500-4
RTECSNumber AH5425000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.