Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60046225 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
35 kg
4 x 2.5 L
6 x 1 L
180 kg
Packaging:
Glass Bottle
Metal Drum
Stainless Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-225 4 x 2.5 L Glass Bottle
60-046-226 6 x 1 L Glass Bottle
60-046-220 180 kg Metal Drum
60-046-221 1 L Glass Bottle
60-046-222 2.5 L Glass Bottle
60-046-224 35 kg Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-046-225 Supplier Solstice Supplier No. 332114X2.5L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Density 0.902g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Odor Like fruit
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 73mmHg (20°C)
Linear Formula CH3COOC2H5
UN Number UN1173
Beilstein 506104
Boiling Point 77°C
Solubility Information ∽90g/L
Formula Weight 88.11g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/ISO/Reagent Ph. Eur./puriss. p.a.
Chemical Name or Material Ethyl acetate
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation Class 3 - PG 2 - Ethyl acetate
RTECSNumber 205-500-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.