Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl acetopyruvate, 98%

Catalog No. AAA1380806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1380806 5 g
AAA1380814 25 g
AAA1380822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1380806 Supplier Thermo Scientific Chemicals Supplier No. A1380806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 615-79-2 | C7H10O4 | 158.15 g/mol

Ethyl acetopyruvate is used in the preparation of 5-methyl-isoxazole-3-carboxylic acid ethyl ester. It is involved in the preparation of ethyl 5-amino-2-methylpyndine-4-carboxyIate by reacting with nitroacetamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetopyruvate is used in the preparation of 5-methyl-isoxazole-3-carboxylic acid ethyl ester. It is involved in the preparation of ethyl 5-amino-2-methylpyndine-4-carboxyIate by reacting with nitroacetamide.

Solubility
Miscible with water, ether,chloroform, methanol and ethanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-79-2
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00009124
InChI Key OYQVQWIASIXXRT-UHFFFAOYSA-N
PubChem CID 69208
IUPAC Name ethyl 2,4-dioxopentanoate
SMILES CCOC(=O)C(=O)CC(C)=O

Specifications

Melting Point 16°C to 18°C
Density 1.126
Boiling Point 108°C to 109°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.473
Quantity 5 g
Beilstein 607062
Synonym ethyl 2,4-dioxovalerate, ethyl acetopyruvate, pentanoic acid, 2,4-dioxo-, ethyl ester, ethyl2,4-dioxovalerate, 2,4-dioxo-pentanoic acid ethyl ester, 2,4-dioxopentanoic acid ethyl ester, 2,4-dioxo-pentanoic-acid-ethyl-ester, 2,4-dioxo-pentanoicacid ethylester, ethyl 2,4-dioxo-valerate, ethyl 2,4-dioxo-pentanoate
Solubility Information Miscible with water,ether,chloroform,methanol and ethanol.
Formula Weight 158.15
Percent Purity 98%
Chemical Name or Material Ethyl acetopyruvate
Show More Show Less
EINECSNumber 210-447-5
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.