Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 190 Proof, FCC, 94.9%, Spectrum™ Chemical
SDP

Catalog No. 18602520 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-520 4 x 4 L Amber Glass Bottle
18-602-519 4 L Amber Glass Bottle
18-602-512 4 L Poly Bottle
18-602-513 4 x 4 L Poly Bottle
18-602-514 20 L Poly Drum
18-602-521 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-602-520 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E10204LTGLCS4
Add to Cart
Add to Cart

E1020, 64-17-5, C2H5OH

Spectrum™ Chemical Ethyl Alcohol, 190 Proof, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Acidity 0.003%
Alkalinity 3 mg/kg
Quantity 4 x 4 L
Packaging Amber Glass Bottle
UN Number UN1170
Concentration 95%
Boiling Point 79°C
Percent Purity 94.9%
Grade FCC
Chemical Name or Material Ethyl Alcohol
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.