Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, 190 Proof, Reagent, ACS, 95%, Spectrum™ Chemical
SDP

Catalog No. 18607997 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Drum
Glass Bottle
Plastic Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-997 4 L Glass Bottle
18-607-995 4 L Plastic Bottle
18-607-998 4 x 4 L Glass Bottle
18-607-996 4 x 4 L Plastic Bottle
18-607-999 20 L Drum
18-608-000 200 L Drum
18-607-990 100 mL Glass Bottle
18-607-991 500 mL Glass Bottle
18-607-993 1 L Glass Bottle
18-607-992 6 x 500 mL Glass Bottle
18-607-994 6 x 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-607-997 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E10294LTGL
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
CAS Max % 0.95, 0.05
Appearance Clear
Quantity 4 L
Flash Point 17°C (63°F)
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.8 at 20°C
Concentration 95%
Boiling Point 79°C
Percent Purity 95%
Grade Reagent
Chemical Name or Material Ethyl Alcohol
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.