Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol 95% Denatured, 100 mL, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S001026 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001026 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001026 Supplier Aldon Corporation Supplier No. IS35034
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-63-0 , 67-56-1 , 64-17-5 , 7732-18-5
Molecular Weight (g/mol) Mixture

Specifications

Quantity 100 mL
Packaging HDPE
Boiling Point 78°C
Percent Purity 100%
Grade Reagent
Chemical Name or Material Ethyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.