Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ethyl Alcohol, 95% Denatured w/IPA and Methanol
SCI_ED

Catalog No. S25309D Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
95% Denatured w/IPA and Methanol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25309D 500 mL 95% Denatured w/IPA and Methanol
S25309E 4 L 95% Denatured w/IPA and Methanol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25309D
Add to Cart
Add to Cart

Denatured Methyl and Isopropyl Alcohol (IPA) solution for educational use. Ideal for classroom labs, cleaning, and demonstrations.

  • Denatured Methyl & IPA
  • Product is for educational use
  • Available in 500 mL and 4 L sizes

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Color Colorless
Density ∽0.8g/mL
Quantity 500 mL
Packaging 95% Denatured w/IPA and Methanol
Physical Form Liquid
Linear Formula C2H5OH
Formula Weight 46.07
Chemical Name or Material Ethyl Alcohol
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.