Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18602525 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Poly Drum
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-525 1 L Amber Glass Bottle
18-602-523 500 mL Amber Glass Bottle
18-602-524 6 x 500 mL Amber Glass Bottle
18-602-526 4 L Amber Glass Bottle
18-602-527 4 L Poly Bottle
18-602-528 6 x 1 L Amber Glass Bottle
18-602-529 4 x 4 L Amber Glass Bottle
18-602-530 4 x 4 L Poly Bottle
18-602-531 20 L Poly Drum
18-602-532 200 L Drum
18-602-522 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18-602-525 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E10281LTGL
Add to Cart
Add to Cart

E1028, 64-17-5, C2H5OH

Spectrum™ Chemical Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS is a pure, 99.5% minimum assay, undenatured ethyl alcohol (ethanol) that is ideal for a variety of research, industrial and educational uses. Spectrum certifies that the Reagent ACS grade is perfect for general laboratory use and offers a solvent that will yield repetitive, consistent results.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 100%
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
Titratable Acid 0.0005 meq/g
Titratable Base 0.0002 meq/g
UN Number UN1170
Concentration 100%
Boiling Point 78°C to 79°C
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Ethyl Alcohol
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.