Learn More
Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Ethyl Alcohol, Absolute, 200 Proof, Reagent, ACS is a pure, 99.5% minimum assay, undenatured ethyl alcohol (ethanol) that is ideal for a variety of research, industrial and educational uses. Spectrum certifies that the Reagent ACS grade is perfect for general laboratory use and offers a solvent that will yield repetitive, consistent results.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 6 x 1 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 0.001% |
| Titratable Acid | 0.0005 meq/g |
| Titratable Base | 0.0002 meq/g |
| UN Number | UN1170 |
| Concentration | 100% |
| Boiling Point | 78°C to 79°C |
| Percent Purity | 99.5% |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.