Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Alcohol Denatured (Proprietary Solvent), Certified, Fisher Chemical™ PROMO

Catalog No. A407P4S24 Shop All Fisherbrand Products
Change view
Click to view available options
Ethyl Alcohol; Packaging: Poly Bottle; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A407P4S24 Ethyl Alcohol; Packaging: Poly Bottle; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A407P4S24

Synonym: Ethanol, Grain Alcohol, Denatured Alcohol, 1-Hydroxyethane

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Color Colorless
Density 0.7905 g/cm3
Quantity 4 L
Flash Point 13.9°C (57°F)
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.7905
Vapor Pressure 40.9 mmHg at 20°C
UN Number 1170
Solubility Information Soluble in Water
Boiling Point 78.5°C
Grade Certified
Chemical Name or Material Ethyl Alcohol, Denatured, Proprietary Solvent
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement Flammable liquid and vapor. Toxic to aquatic life.
Precautionary Statement Toxic if inhaled. May damage fertility or the unborn child.
DOTInformation DOT Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.