Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Arachidate 95.0+%, TCI America™
SDP

Catalog No. A089925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A089925G 25 g
A08995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A089925G Supplier TCI America Supplier No. A089925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18281-05-5
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.59
MDL Number MFCD00056233
InChI Key YBKSMWBLSBAFBQ-UHFFFAOYSA-N
Synonym Icosanoic Acid Ethyl Ester, Arachidic Acid Ethyl Ester, Ethyl Icosanoate
PubChem CID 29009
ChEBI CHEBI:84859
IUPAC Name ethyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 340.59
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Arachidate
EINECSNumber (2)-0798
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.