Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Benzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. B0032500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0032500G 500 g
B003225G 25 g
B0032100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0032500G Supplier TCI America Supplier No. B0032500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 515-46-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.225
MDL Number MFCD00025033
InChI Key XDRMBCMMABGNMM-UHFFFAOYSA-N
Synonym Benzenesulfonic Acid Ethyl Ester
PubChem CID 10585
IUPAC Name ethyl benzenesulfonate
SMILES CCOS(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 156°C
Quantity 500 g
Formula Weight 186.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.