Learn More
Sigma Organic Chemistry Ethyl benzoate | 500G | 93-89-0 | MFCD00009109
Supplier: Sigma Organic Chemistry E12907500G

Ethyl benzoate, 500G
About This Item:
Synonym(s): Benzoic acid ethyl ester
Linear Formula: C6H5COOC2H5
Molecular Weight: 150.17
CAS Number: 93-89-0
MDL Number: MFCD00009109
UNSPSC Code: 12352100
Purity: >=99%
Boiling Point: 212 C (lit.)
Melting Point: -34 C (lit.)
Density: 1.045 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.504 (lit.)
Storage: room temp
EINECS Number: 202-284-3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.