Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl benzoylacetate

Catalog No. 281492500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-149-2500ML 500 mL
28-149-2100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-149-2500ML Supplier Sigma Aldrich Supplier No. 281492500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl benzoylacetate is an ester. it undergoes microbial reduction by bakersα yeast (saccharomyces cerevisiae), beauveria sulfurescens or geotrichum candidum to afford ethyl (s)-3-hydroxy-3-phenylpropionate. it undergoes claisen condensation reaction with malononitrile to afford 2-cyano-5-phenyl-3,5-dioxopentanonitrile which on cyclization followed by coupling with diazonium salts yields azo derivatives.ethyl benzoylacetate was sed in the preparation of:
  • Triazipinones
  • Ethyl 2-fluoro-2-benzolyacetate
  • 1,2,4-triazolo[1,5-a]pyrimidine antibiotic essramycin
  • Benzo[c]furan-4,7-diones
  • Iodonium ylides

Specifications

CAS 94-02-0
Density 1.11 g/mL (at 25°C (literature))
Boiling Point 265°C to 270°C (lit.)
Molecular Formula C11H12O3
Refractive Index n20/D 1.52 (literature)
Linear Formula C6H5COCH2COOC2H5
MDL Number MFCD00009196
Quantity 500 mL
Synonym Benzoylacetic acid ethyl ester; Ethyl 3-oxo-3-phenylpropionate
Molecular Weight (g/mol) 192.21
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.