Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl benzoylacetate, 90+%

Catalog No. AAL050300B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL050300B 1000 g
AAL0503018 50 g
AAL0503030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL050300B Supplier Thermo Scientific Chemicals Supplier No. L050300B
Add to Cart

CAS: 94-02-0 | C11H12O3 | 192.214 g/mol

Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

Solubility
Miscible with ethanol and ether. Immiscible with water.

Notes
Air and light sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-02-0
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00009196
InChI Key GKKZMYDNDDMXSE-UHFFFAOYSA-N
Synonym ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate
PubChem CID 7170
IUPAC Name ethyl 3-oxo-3-phenylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1

Specifications

Density 1.113
Boiling Point 266°C to 270°C
Flash Point 140°C (284°F)
Refractive Index 1.529
Quantity 1000 g
Beilstein 389944
Merck Index 14,3767
Solubility Information Miscible with ethanol and ether. Immiscible with water.
Formula Weight 192.21
Percent Purity 96%
Chemical Name or Material Ethyl benzoylacetate
Show More Show Less
EINECSNumber 202-295-3
RTECSNumber AF4878000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.