Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (Benzyloxy)acetate 98.0+%, TCI America™
SDP

Catalog No. E12251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12251G 1 g
E12255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12251G Supplier TCI America Supplier No. E12251G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32122-09-1
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD06204614
InChI Key PHSWWKXTGAJPCQ-UHFFFAOYSA-N
Synonym (Benzyloxy)acetic Acid Ethyl Ester
PubChem CID 290299
IUPAC Name ethyl 2-phenylmethoxyacetate
SMILES CCOC(=O)COCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 287°C
Quantity 1 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl (Benzyloxy)acetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.