Learn More
Sigma Organic Chemistry Ethyl butyrate | 1L | 105-54-4 | MFCD00009394
Supplier: Sigma Organic Chemistry E157011L

Ethyl butyrate, 1L
About This Item:
Synonym(s): Ethyl butanoate; Butyric acid ethyl ester
Linear Formula: CH3CH2CH2C(O)OC2H5
Molecular Weight: 116.16
CAS Number: 105-54-4
MDL Number: MFCD00009394
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 120 C (lit.)
Melting Point: -93 C (lit.)
Density: 0.875 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.392 (lit.)
Storage: room temp
EINECS Number: 203-306-4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.