Learn More
Ethyl chloroacetate, 99%
Description
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl chloroacetate is a reagent used in the preparation of 5 member heterocycles. It is used as pharmaceutical and organic intermediate. It is used as a solvent for organic synthesis.
Solubility
Soluble in water 20 g/L (20°C).
Notes
Incompatible with acids, bases, oxidizing agents, reducing agents.
Specifications
Specifications
| Melting Point | -26°C |
| Density | 1.159 |
| Boiling Point | 142°C to 144°C |
| Flash Point | 54°C (149°F) |
| Odor | Pungent |
| Refractive Index | 1.421 |
| Linear Formula | ClCH2CO2CH2CH3 |
| Quantity | 2500 g |
| UN Number | UN1181 |
| Beilstein | 506455 |
| Show More |
Safety and Handling
| Hazard Category | H226-H300-H311+H331-H315-H317-H318 |
| Hazard Statement | GHS H Statement H301-H311-H331-H226 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ETHYL CHLOROACETATE |
| EINECSNumber | 203-294-0 |
| RTECSNumber | AF9110000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.