Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Chlorofluoroacetate 98.0+%, TCI America™
SDP

Catalog No. C146725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C146725G 25 g
C14675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C146725G Supplier TCI America Supplier No. C146725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 401-56-9
Molecular Formula C4H6ClFO2
Molecular Weight (g/mol) 140.538
MDL Number MFCD00039333
InChI Key WUHVJSONZHSDFC-UHFFFAOYSA-N
Synonym ethyl chlorofluoroacetate, acetic acid, chlorofluoro-, ethyl ester, chlorofluoroacetic acid ethyl ester, ethyl chloro fluoro acetate, ethyl fluorochloroacetate, acetic acid, 2-chloro-2-fluoro-, ethyl ester, acmc-1csjg, ksc492m6l, ethyl 2-chloro-2-fluoro-acetate
PubChem CID 101668
IUPAC Name ethyl 2-chloro-2-fluoroacetate
SMILES CCOC(=O)C(F)Cl

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 g
UN Number 2924
Formula Weight 140.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Chlorofluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.