Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cis-9-Hexadecenoate 95.0+%, TCI America™
SDP

Catalog No. H05011G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05011G 1 g
H0501100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05011G Supplier TCI America Supplier No. H05011G

Chemical Identifiers

CAS 56219-10-4
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00056314
InChI Key JELGPLUONQGOHF-KTKRTIGZSA-N
Synonym Ethyl Palmitoleate, cis-9-Hexadecenoic Acid Ethyl Ester, Palmitoleic Acid Ethyl Ester
PubChem CID 6436624
ChEBI CHEBI:84934
IUPAC Name ethyl (Z)-hexadec-9-enoate
SMILES CCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 1 g
Formula Weight 282.47
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl cis-9-Hexadecenoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.