Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl cyanoformate, 99%

Catalog No. AAL1577714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1577714 25 g
AAL1577706 5 g
AAL1577722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1577714 Supplier Thermo Scientific Chemicals Supplier No. L1577714
Add to Cart

CAS: 623-49-4 | C4H5NO2 | 99.09 g/mol

Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl cyanoformate is used as reagent in the preparation of N-substituted amindinoformic acid and ethyl-4-quinazoline -2-carboxylate.

Solubility
Miscible with chloroform and methanol.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-49-4
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001836
InChI Key MSMGXWFHBSCQFB-UHFFFAOYSA-N
Synonym carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
PubChem CID 69336
IUPAC Name ethyl cyanoformate
SMILES CCOC(=O)C#N

Specifications

Density 1.003
Boiling Point 115°C to 116°C
Flash Point 24°C (75°F)
Refractive Index 1.382
Quantity 25 g
UN Number UN3275
Beilstein 635955
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Miscible with chloroform and methanol.
Formula Weight 99.09
Percent Purity 99%
Chemical Name or Material Ethyl cyanoformate
Show More Show Less
Hazard Category H226-H301+H311+H331
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 210-797-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.