Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cyanoformate 98.0+%, TCI America™
SDP

Catalog No. C126625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C126625G 25 g
C12665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C126625G Supplier TCI America Supplier No. C126625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-49-4
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001836
InChI Key MSMGXWFHBSCQFB-UHFFFAOYSA-N
Synonym carbonocyanidic acid, ethyl ester, ethyl carbonocyanidate, 2-ethoxy-2-oxoacetonitrile, cyanoformic acid ethyl ester, ethyl carbonocyanidoate, formic acid, cyano-, ethyl ester, cyanocarbonyloxyethane, ethylcyanidocarbonat, ethyl cyanoformate, ethyl cyanocarbonate
PubChem CID 69336
IUPAC Name ethyl cyanoformate
SMILES CCOC(=O)C#N

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 g
UN Number 3275
Formula Weight 99.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyanoformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.