Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Cyclohexylacetate 98.0+%, TCI America™
SDP

Catalog No. C107625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C107625ML 25 mL
C1076500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C107625ML Supplier TCI America Supplier No. C107625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5452-75-5
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00001517
InChI Key ZBDAMDWKXGTKBT-UHFFFAOYSA-N
Synonym ethyl cyclohexylacetate, ethyl cyclohexaneacetate, cyclohexaneacetic acid, ethyl ester, ethyl-2-cyclohexylacetate, ethylcyclohexylacetate, unii-v36o028g9r, cyclohexylacetic acid ethyl ester, cyclohexaneacetic acid ethyl ester, cyclohexyl acetic acid ethyl ester, pubchem20245
PubChem CID 21595
IUPAC Name ethyl 2-cyclohexylacetate
SMILES CCOC(=O)CC1CCCCC1

Specifications

Color Colorless
Boiling Point 212°C
Quantity 25 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Cyclohexylacetate
RTECSNumber GU6555000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.