Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl D-(-)-Mandelate 98.0+%, TCI America™
SDP

Catalog No. M134425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M134425G 25 g
M13445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M134425G Supplier TCI America Supplier No. M134425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10606-72-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00064248
InChI Key SAXHIDRUJXPDOD-SECBINFHSA-N
Synonym D-(-)-Mandelic Acid Ethyl Ester
PubChem CID 6951556
ChEBI CHEBI:78406
IUPAC Name ethyl (2R)-2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Ethyl D-(-)-Mandelate
EINECSNumber (3)-3841
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.