Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Diacetoacetate 96.0+%, TCI America™
SDP

Catalog No. D217025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D217025G 25 g
D21705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D217025G Supplier TCI America Supplier No. D217025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 603-69-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009864
InChI Key YMCDYRGMTRCAPZ-UHFFFAOYSA-N
PubChem CID 79063
IUPAC Name ethyl 2-acetyl-3-oxobutanoate
SMILES CCOC(=O)C(C(=O)C)C(=O)C

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 g
Synonym ethyl diacetoacetate, ethyl 2-acetylacetoacetate, butanoic acid, 2-acetyl-3-oxo-, ethyl ester, 3-ethoxycarbonylacetylacetone, ethyl diacetylacetate, ethyl diacetyl acetate, ethyl diaceto acetate, 2-acetyl-3-oxo-butyric acid ethyl ester, butanoic acid,2-acetyl-3-oxo-, ethyl ester, ethyldiacetoacetate
Formula Weight 172.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Diacetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.