Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, contains 2% ethanol, >99.9% (excluding ethanol), Solstice

Catalog No. 6000598 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-98 4 x 4 L Poly-coated Bottle
60-005-97 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-98 Supplier Solstice Supplier No. AH1064PC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-29-7 , 64-17-5
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

CAS Max % 98.0000%, 2.0000%
Density 0.713 g/cm3
Quantity 4 x 4 L
Flash Point −45°C
Packaging Poly-coated Bottle
Water 0.03% max.
UN Number UN1155
Beilstein 1696894
Boiling Point 35°C
Solubility Information 689% in Water at 20°C
Formula Weight 74.12
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Ethyl Ether, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, contains 2% ethanol, >99.9% (excluding ethanol)
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.