Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ethyl Ether, Anhydrous, ACS Grade
SCI_ED

Catalog No. S25903 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25903 4 L
S25903L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25903
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

High-purity ethyl ether anhydrous, ACS grade, ideal for chemistry instruction and laboratory experiments. Supplied in 4 L quantity for educational use.

  • Chemical Name: Ethyl Ether, Anhydrous
  • Grade: ACS (American Chemical Society)
  • Purity: High-grade suitable for educational and laboratory experiments
  • Packaging: 4-liter container
  • Color: Colorless liquid
  • Formula Weight: 74.12 g/mol
  • Physical Form: Liquid
  • Intended Use: Educational use only, not for research or clinical applications
  • Certificate of Analysis not provided with this product

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Color Colorless
Quantity 4 L
Packaging ACS Grade
Physical Form Liquid
Formula Weight 74.12
Chemical Name or Material Ethyl Ether Anhydrous
Safety handling Check Alert storage code Red
Hazard Category Flammable
ShelfLife Two years
DOTInformation DOT Class 3, I: Flammable, 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.