Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, Anhydrous, GR, ACS, MilliporeSigma™

Catalog No. mex01908 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Tinplate Metal Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MEX01908 500 mL Tinplate Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MEX01908 Supplier MilliporeSigma Supplier No. EX01908

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116.3°C
Quantity 500 mL
Flash Point -40°C
Packaging Tinplate Metal Can
Physical Form Clear liquid
Residue after Evaporation 0.001%
Titratable Acid 0.0002meq/g
Vapor Pressure 587 hPa (20°C)
Water 0.03%
Solubility Information 69 g/L
Formula Weight 74.12
Percent Purity 99.0%
Show More Show Less
EINECSNumber 200-467-2
Recommended Storage Store at +2°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.