Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%, Honeywell™ Burdick & Jackson

Catalog No. 6000535 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-005-35 204 L Metal Drum
60-005-34 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-005-35 Supplier Honeywell Chemicals Supplier No. PP108204

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

CAS Max % 100.0000%, ≤100.0000%
Density 0.713 g/cm3
Quantity 204 L
Flash Point −45°C
Packaging Metal Drum
UN Number UN1155
Beilstein 1696894
Water 0.03% max.
Solubility Information 689% in Water at 20°C
Boiling Point 35°C
Formula Weight 74.12
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Ethyl Ether, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, contains BHT, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: I; Proper Shipping Name: Diethyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.